Sample tubes
1,5 ml, Free standing, Clear, with side label, PP
Catalog # BZYM710020
Marketsource
Supplier: BIOZYM
Jump to:
Product Details & Documents
PP, translucent.
- Excellent chemical resistance
- Excellent temperature resistance
With or without screw caps.
Specifications
- Capacity:1,5 ml
- Colour:Clear
- Description:with side label
- Material:PP
- Seq:07
- Base:Free standing