Sample tubes
0,5 ml, Free standing, Amber, with side label, PP
BZYM710016
Marketsource
: BIOZYM
PP, translucent.
- Excellent chemical resistance
- Excellent temperature resistance
With or without screw caps.
- Capacity:0,5 ml
- Colour:Amber
- Description:with side label
- Material:PP
- Seq:04
- Base:Free standing