Order Entry
France
Orders LinkContactUsLinkComponent
Sample tubes
Sample tubes
0,5 ml, Free standing, Clear, with side label, PP
Catalog # BZYM710015
Marketsource
Supplier:   BIOZYM
Sample tubes
Catalog # BZYM710015
Supplier:  BIOZYM
Marketsource
Sample tubes
0,5 ml, Free standing, Clear, with side label, PP
Price
Inquire For Price
Log in for availability
1 * 500 Pce
Product Details & Documents
PP, translucent.
  • Excellent chemical resistance
  • Excellent temperature resistance
With or without screw caps.
Specifications
  • Capacity:
    0,5 ml
  • Colour:
    Clear
  • Description:
    with side label
  • Material:
    PP
  • Seq:
    03
  • Base:
    Free standing