Sample tubes
2,0 ml, Conical bottom, Clear, with side label, PP
Catalog # BZYM710011
Marketsource
Supplier: BIOZYM
Jump to:
Product Details & Documents
PP, translucent.
- Excellent chemical resistance
- Excellent temperature resistance
With or without screw caps.
Specifications
- Capacity:2,0 ml
- Colour:Clear
- Description:with side label
- Material:PP
- Seq:09
- Base:Conical bottom