Order Entry
France
Orders LinkContactUsLinkComponent
Process wipes, KIMTECH PREP*
Process wipes, KIMTECH PREP*
Roll, Blue, 380×340 mm, 1 roll of 500 sheets
Catalog # 115-2222
Supplier:   KIMBERLY CLARK
Process wipes, KIMTECH PREP*
Catalog # 115-2222
Supplier:  KIMBERLY CLARK
Process wipes, KIMTECH PREP*
Process wipes, KIMTECH PREP*
Roll, Blue, 380×340 mm, 1 roll of 500 sheets
Price
439,00 €
Log in for availability
1 * 1 ROU
Product Details & Documents
Strong, low-lint wipes for repeat use cleaning with chemicals and solvents. Perfect for printing, pre and post-paint applications, glass and appliance manufacturing, engine maintenance.
  • PP, reusable
  • Great for wiping oils and grease
  • Acid, base, and solvent resistant
  • Roll 115-2222 is compatible with dispensers 115-0045, 115-1533 and 115-2038
Colour: Blue
Specifications
  • Packed:
    1 roll of 500 sheets
  • Colour:
    Blue
  • Description:
    Roll
  • L×W:
    380×340 mm