Jump to:
Product Details & Documents
Synonyms: Methylrosanilide chloride, Basic violet 3, Viocid, N-(4-(Bis(4-(dimethylamino)phenyl)methylene)cyclohexa-2,5-dien-1-ylidene)-N-methylmethanamonium chloride, Violet crystal, Methyl violet 10B, Dimethyl((di(4-(dimethylamino)phenyl)methylen)-2,5-cyclohexadien-1-ylidene)ammonium chloride, CI 42555, 4-[4,4'-Bis(dimethylamino) benzhydrylidene]cyclohexa-2,5-dien-1-ylidene]dimethylammonium chloride, Hexamethyl violet, Hexamethyl-p-rosaniline chloride, Vianin, Gentian violet
- CAS Number:548-62-9
- Formula:C₂₅H₃₀ClN₃
- MDL Number:MFCD00011750
- Molecular Weight:407.99 g/mol
- Merck Index:13,04406
- Melting Pt:189…194 °C
- Density:1.19 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:208-953-6
- UN:3077
- ADR:9,III
- :M-Clarity™ quality level = MQ200
Specifications
Specification Test Results
- Solubility:10 g/l (20 °C)
- Melting Point:189 - 194 °C
- Molar Mass:407.99 g/mol
- Bulk Density:220 - 400 kg/m3
- Density:1.19 g/cm3 (20 °C)
- pH:2.5 - 3.5 (10 g/l, H2O, 20 °C)
- Hazard Class:9
- Hazard Class Description:Miscellaneous Hazardous Material
Specifications
- Pack type:Glass bottle
- Pk:1 kg



