Order Entry
France
Orders LinkContactUsLinkComponent
Solution 192

Solution 192

Plastic drum, 20 L
Catalog # PRLSMC311897.9020
Flammable
Irritant
Supplier:   PROLABO CMP
Solution 192
Catalog # PRLSMC311897.9020
Supplier:  PROLABO CMP
CAS Number:  
Solution 192

Plastic drum, 20 L

CAS Number:  
Price
320,00 €
Log in for availability
1 * 20 L
Product Details & Documents

Hydroxylamine sulphate 2 g + eriochrome black 0,5 g + Isopropanol 300 ml + water QSP 1 000 ml

  • Melting Pt:-89 °C
  • UN:1993
  • ADR:3,II
Specifications

Specifications

  • Pack type:
    Plastic drum
  • Pk:
    20 L

Specifications