0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
France
Orders Link
ContactUsLinkComponent
Sodium hydroxide
Sodium hydroxide 10% in aqueous solution
Sodium hydroxide 10% in aqueous solution
Plastic container, 10 L
PRLSMC302652.9010
:
PROLABO CMP
Sodium hydroxide 10% in aqueous solution
PRLSMC302652.9010
:
PROLABO CMP
:
1310-73-2
Sodium hydroxide 10% in aqueous solution
Plastic container, 10 L
:
1310-73-2
272,00 €
1 * 10 L
-
+
:
Caustic soda
:
1310-73-2
:
NaOH
:
MFCD00003548
:
40 g/mol
:
119…135 °C (1013 hPa)
:
9…10 °C
:
1,53 g/cm³ (20 °C)
:
Ambient
:
215-185-5
:
1824
:
8,II
Pack type:
Plastic container
Pk:
10 L
VWR®, Qualitative Filter Papers
#: 516-0809
: VWR Collection
Hydrochloric acid 4 mol/l (4 N), VOLUSOL® volumetric solution
#: 310701.5000
: VWR Chemicals
Petroleum spirit, 40…60 °C, GPR RECTAPUR®
#: 23826.360
: VWR Chemicals