Order Entry
France
Orders LinkContactUsLinkComponent
Sodium hydroxide 10% in aqueous solution

Sodium hydroxide 10% in aqueous solution

Plastic bottle, 5 L
Catalog # PRLSMC302652.5000
Corrosive
Supplier:   PROLABO CMP
Sodium hydroxide 10% in aqueous solution
Catalog # PRLSMC302652.5000
Supplier:  PROLABO CMP
CAS Number:  1310-73-2
Sodium hydroxide 10% in aqueous solution

Plastic bottle, 5 L

CAS Number:  1310-73-2
Price
123,00 €
Log in for availability
1 * 5 L
Product Details & Documents
Synonyms:  Caustic soda
  • CAS Number:1310-73-2
  • Formula:NaOH
  • MDL Number:MFCD00003548
  • Molecular Weight:40 g/mol
  • Boiling Pt:119…135 °C (1013 hPa)
  • Melting Pt:9…10 °C
  • Density:1,53 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:215-185-5
  • UN:1824
  • ADR:8,II

Documents

Specifications

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    5 L

Specifications