Order Entry
France
Orders LinkContactUsLinkComponent
Sodium hydroxide 10% in aqueous solution

Sodium hydroxide 10% in aqueous solution

Plastic bottle, 1 L
Catalog # PRLSMC302652.1000
Corrosive
Supplier:   PROLABO CMP
Sodium hydroxide 10% in aqueous solution
Catalog # PRLSMC302652.1000
Supplier:  PROLABO CMP
CAS Number:  1310-73-2
Sodium hydroxide 10% in aqueous solution

Plastic bottle, 1 L

CAS Number:  1310-73-2
Price
37,90 €
Log in for availability
1 * 1 L
Product Details & Documents
Synonyms:  Caustic soda
  • CAS Number:1310-73-2
  • Formula:NaOH
  • MDL Number:MFCD00003548
  • Molecular Weight:40 g/mol
  • Boiling Pt:119…135 °C (1013 hPa)
  • Melting Pt:9…10 °C
  • Density:1,53 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:215-185-5
  • UN:1824
  • ADR:8,II

Documents

Specifications

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 L

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.