Order Entry
France
Orders LinkContactUsLinkComponent
Dynamometers
Dynamometers
LG-10KG, 0 - 10 kgf, 0,05 kgf
Catalog # 619-0248
Supplier:   LLOYD INSTRUMENTS
Dynamometers
Catalog # 619-0248
Supplier:  LLOYD INSTRUMENTS
Dynamometers
LG-10KG, 0 - 10 kgf, 0,05 kgf
Price
986,88 €
Log in for availability
1 * 1 Pce
Product Details & Documents
The Chatillon® LG series mechanical force gauge is ideal for a wide range of force testing applications. Features a larger dial for improved resolution and readability, peak hold button, ergonomic shape and improved accuracy of +0,5% of full scale.
  • Graduation styles : KGF and Newtons
  • Excellent, economical choice for tensile or compression testing
  • NIST traceable calibration certificate
Specifications
  • Range:
    0 - 10 kgf
  • Description:
    LG-10KG
  • Seq:
    04
  • Readability:
    0,05 kgf