Order Entry
France
Orders LinkContactUsLinkComponent
Dynamometers
LG-3KG, 0 - 3 kgf, 0,015 kgf
  619-0254
Dynamometers
  619-0254
Dynamometers
LG-3KG, 0 - 3 kgf, 0,015 kgf
1 * 1 Pce
The Chatillon® LG series mechanical force gauge is ideal for a wide range of force testing applications. Features a larger dial for improved resolution and readability, peak hold button, ergonomic shape and improved accuracy of +0,5% of full scale.
  • Graduation styles : KGF and Newtons
  • Excellent, economical choice for tensile or compression testing
  • NIST traceable calibration certificate
  • Range:
    0 - 3 kgf
  • Description:
    LG-3KG
  • Seq:
    02
  • Readability:
    0,015 kgf