Order Entry
France
Orders LinkContactUsLinkComponent
Ammonia solution 25% 100 ml/l solution

Ammonia solution 25% 100 ml/l solution

Plastic bottle, 2,5 L
Catalog # PRLS85163.320
Irritant
Environment
Supplier:   PROLABO CMP
Ammonia solution 25% 100 ml/l solution
Catalog # PRLS85163.320
Supplier:  PROLABO CMP
CAS Number:  1336-21-6
Ammonia solution 25% 100 ml/l solution

Plastic bottle, 2,5 L

CAS Number:  1336-21-6
Price
60,30 €
Log in for availability
1 * 2,5 L
Product Details & Documents

Concentrated solution to dilute to 100 mL/L.

Synonyms:  Ammonia aqueous, Ammonium hydroxide solution, Ammonium hydroxide, Ammonia water
  • CAS Number:1336-21-6
  • Formula:NH₃
  • MDL Number:MFCD00066650
  • Molecular Weight:35.05 g/mol
  • Density:0.92 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:215-647-6
  • ADR:8,III
Specifications

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    2,5 L

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.