Order Entry
France
Orders LinkContactUsLinkComponent
Micro pipettes, disposable, calibrated
50 µl, Green
  DRUM2-000-050
 :   DRUMMOND
Micro pipettes, disposable, calibrated
  DRUM2-000-050
 :  DRUMMOND
Micro pipettes, disposable, calibrated
50 µl, Green
113,00 €
1 * 250 Pce
These precision calibrated micro pipettes are available in sizes ranging from 1 to 200 µl. Ideal for a variety of uses in liquid transfer work.
  • Fire polished
  • Accuracy of ±0,5%
  • Colour coded micro pipettes with sizes imprinted
  • Anticoagulant treatment for clinical applications (special limited sizes)
 : 250 colour coded micro pipettes, plus one 15" aspirator tube, are supplied in a conveniently designed, easy to store cylinder.
  • Colour code:
    Green
  • Capacity:
    50 µl
Accessories
  •  
    Description
  • DRUM2-000-000
    Aspirator tube assembly, 38 cm
    1 * 1 Pce - 8,10 €