Spring scissors, Noyes
Straight, blunt/blunt, 120 mm
Catalog # SVLM37.060.12
Supplier: SERVICE LABO MEDICAL
Jump to:
Product Details & Documents
German stainless steel, with satin finish. These spring scissors are ideal for surgical applications.
Specifications
- Type:Straight, blunt/blunt
- Length:120 mm
- Seq:01