Order Entry
France
Orders LinkContactUsLinkComponent
Fire blankets, Range A
Fire blanket, for wall-mounting, box dimensions: 160×35×285 mm, 1 kg, 1200×1200 mm
  TRIO120120RRP
 :   TRIONYX
Fire blankets, Range A
  TRIO120120RRP
 :  TRIONYX
Fire blankets, Range A
Fire blankets, Range A
Fire blanket, for wall-mounting, box dimensions: 160×35×285 mm, 1 kg, 1200×1200 mm
122,57 €
1 * 1 Pce
These fire blankets made of fibreglass (420 g/m²) are intended for wall-mounting. They resist at least 900 °C temperature.
  • Wall fastening allows immediate sight and rapid access
  • Extra-flat, very light and resistant
  • Immediate opening with two tapes
  • Instruction manual affixed on the blanket
Fixed on the wall the fire blanket is supplied with two instantaneous release tapes which function as handles when the blanket is pulled from the container. The blanket itself acts as a heat shield protecting the users from the fire. When draped over a fire, it smothers the fire by starving it of oxygen, leading to rapid extinction. The fire blanket can also be used to wrap around a person whose clothes are on fire or to wrap around a hot or burning object.
 : EN 1869-1997
  • Description:
    Fire blanket, for wall-mounting, box dimensions: 160×35×285 mm, 1 kg
  • L×W:
    1200×1200 mm
  • Seq:
    01