Order Entry
France
Orders LinkContactUsLinkComponent
Taps
Taps
Demineralised water gooseneck tap, for font, On backsplash*
Catalog # 132-2710
Supplier:   EQUIP LABO
Taps
Catalog # 132-2710
Supplier:  EQUIP LABO
Taps
Taps
Demineralised water gooseneck tap, for font, On backsplash*
Price
201,00 €
Log in for availability
1 * 1 Pce
Product Details & Documents
The taps are of 3 types:
  • Taps for hard and demineralised water
  • Taps for combustible gases
  • Taps for air, compressed gas and vacuum
Straight hose length 80 cm, exterior stainless steel braid, interior neoprene sheath. Suitable for taps on the table-top (for cold water, hot water, compressed air). Outlet by 15/21 female nut for connection on 15/21 (¹/₂") male standby gas feed.
Elbow hose with same characteristics as straight hoses but adapted to plumbing fittings on splash back (for cold water, hot water, compressed air).
Ordering information: Different sockets (individual and in a row) are available on request.
Caution: * All of these valves are available in a counter-top version.
Specifications
  • Description:
    Demineralised water gooseneck tap, for font, On backsplash*