Order Entry
France
Orders LinkContactUsLinkComponent
Sample vials, clear, with screw thread, KIMBLE®
Sample vials, clear, with screw thread, KIMBLE®
Catalog # SCERC40903000F0C2
Supplier:   DWK Life Sciences
Sample vials, clear, with screw thread, KIMBLE®
Catalog # SCERC40903000F0C2
Supplier:  DWK Life Sciences
Sample vials, clear, with screw thread, KIMBLE®
Sample vials, clear, with screw thread, KIMBLE®
Price
156,00 €
Log in for availability
1 * 100 Pce
Product Details & Documents
Screw threaded sample vials made from soda-lime glass, with white PE caps unattached.