Order Entry
France
Orders LinkContactUsLinkComponent
Disposable cuvettes
Disposable cuvettes
Blue
Catalog # 612-5690
Supplier:   Brand
Disposable cuvettes
Catalog # 612-5690
Supplier:  Brand
Disposable cuvettes
Disposable cuvettes
Blue
Price
16,30 €
Log in for availability
1 * 100 Pce
Product Details & Documents
Disposable cuvettes often replace expensive and sensitive cuvettes made from glass and quartz.
  • High chemical resistance for use with polar solvents, acids and caustic solutions
  • Measurements at 260 or 280 nm and in the visible wavelength range
  • Round coloured caps, for microcuvettes only, ensure easy sample identification and provide a tight seal to –20 °C
Suitable for photometric measurements of proteins, DNA and RNA in the UV range.
 : Individually wrapped micro UV cuvettes are free of DNase, DNA and RNase.
Specifications
  • Description:
    Blue
  • Seq:
    09