Order Entry
France
ContactUsLinkComponent
Vinometer, measuring range:0-25%vol, gives approximate dates of percentage of alcohol in wine, solid stem, white backed, length:130m m. 1 * 1 items
Catalog # AMARH850450
Supplier:  Amarell
Vinometer, measuring range:0-25%vol, gives approximate dates of percentage of alcohol in wine, solid stem, white backed, length:130m m. 1 * 1 items
Catalog # AMARH850450
Supplier:  Amarell
Frequently Bought Together