Order Entry
France
Orders LinkContactUsLinkComponent
Ammonia solution 25% ≥99.99% (metals basis)

Ammonia solution 25% ≥99.99% (metals basis)

Glass bottle, 250 mL
Catalog # 87903.AK
Corrosive
Irritant
Environment
Ammonia solution 25% ≥99.99% (metals basis)
Catalog # 87903.AK
CAS Number:  1336-21-6
Ammonia solution 25% ≥99.99% (metals basis)
Ammonia solution 25% ≥99.99% (metals basis)

Glass bottle, 250 mL

CAS Number:  1336-21-6
Price
182,00 €
Log in for availability
1 * 250 mL
Product Details & Documents

Purity: 99.99% CAS No.: 1336-21-6 Molecular Formula: H5NO Formula Weight: 35.05 Physical Form: Liquid Density: 0.91 Appearance: Colourless Odour: Ammonia-like pH: 12 Merck Reference: 14,494 UN No.: 2672 MDL No.: MFCD00066650

Synonyms:  Ammonia aqueous, Ammonium hydroxide solution, Ammonium hydroxide, Ammonia water
  • CAS Number:1336-21-6
  • Formula:NH₃
  • MDL Number:MFCD00066650
  • Molecular Weight:35.05 g/mol
  • Merck Index:14,494
  • Boiling Pt:32…37.7 °C (1013 hPa)
  • Melting Pt:–60…–57.5 °C
  • Density:0.91
  • Storage Temperature:Ambient
  • EINECS:215-647-6
  • UN:2672
  • ADR:8,III
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    250 mL

Specifications