Nickel plating solution, semi-bright finish
1 L
Supplier: Thermo Fisher Scientific
Nickel plating solution, semi-bright finish
Catalog # 42026.K2
Supplier: Thermo Fisher Scientific
CAS Number:
Jump to:
Product Details & Documents
Physical Form: Liquid Appearance: Green UN No.: UN3287 MDL No.: MFCD00011137
- MDL Number:MFCD00011137
- Storage Temperature:Ambient
- UN:3287
- ADR:6.1,III
Documents
Specifications
Specifications
- Pk:1 L
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



