Nickel plating solution, semi-bright finish
250 mL
Supplier: Thermo Fisher Scientific
Nickel plating solution, semi-bright finish
Catalog # 42026.AK
Supplier: Thermo Fisher Scientific
CAS Number:
Jump to:
Product Details & Documents
Physical Form: Liquid Appearance: Green UN No.: UN3287 MDL No.: MFCD00011137
- MDL Number:MFCD00011137
- Storage Temperature:Ambient
- UN:3287
- ADR:6.1,III
Specifications
Specifications
- Pk:250 mL



