Order Entry
France
Orders LinkContactUsLinkComponent
Push-in caps for tubes
Push-in caps for tubes
LDPE, Caps, Ø 29 mm, Natural
Catalog # 215-0216
Supplier:   SEMADENI
Push-in caps for tubes
Catalog # 215-0216
Supplier:  SEMADENI
Push-in caps for tubes
Push-in caps for tubes
LDPE, Caps, Ø 29 mm, Natural
Price
27,40 €
Log in for availability
1 * 100 Pce
This product has been blocked by your organization. Please contact your purchasing department for more information
Product Details & Documents
LDPE, translucent.
Caps with rim for sealing test tubes, flasks and bottles.
Specifications
  • Colour:
    Natural
  • Description:
    Caps, Ø 29 mm
  • Material:
    LDPE
  • Seq:
    18