Order Entry
France
ContactUsLinkComponent
Container, straight, capacity 180ml, material natural polypropylene, diameter 52mm, height 102mm, color of screw cap red, shapolyeth ylene assembled, CE, IVD, clinical and industrial. 1 * 264 items
Catalog # 216-2598
Supplier:  VWR Collection
Container, straight, capacity 180ml, material natural polypropylene, diameter 52mm, height 102mm, color of screw cap red, shapolyeth ylene assembled, CE, IVD, clinical and industrial. 1 * 264 items
Catalog # 216-2598
Supplier:  VWR Collection
Frequently Bought Together