Order Entry
France
Orders LinkContactUsLinkComponent
Small flanges with hose nozzle
Small flanges with hose nozzle
Small flanges, DN 40, Aluminium, 15 mm
Catalog # 185-2259
Supplier:   VACUUBRAND
Small flanges with hose nozzle
Catalog # 185-2259
Supplier:  VACUUBRAND
Small flanges with hose nozzle
Small flanges with hose nozzle
Small flanges, DN 40, Aluminium, 15 mm
Price
75,30 €
Log in for availability
1 * 1 Pce
Product Details & Documents
These flanges are made from aluminium or plastic.
  • Fast, easy assembly and disassembly to/from pumps
  • Safe, vacuum tight connection with optional centring and clamping rings
  • A wide range of flanges with hose nozzles for various internal hose diameters are available
For connecting pumps to vacuum hoses.
Specifications
  • Tubing int.Ø:
    15 mm
  • Description:
    Small flanges, DN 40
  • Material:
    Aluminium
  • Seq:
    13