Order Entry
France
Orders LinkContactUsLinkComponent
Giemsa's stain solution for microscopy, Sigma-Aldrich®

Giemsa's stain solution for microscopy, Sigma-Aldrich®

Glass bottle, 100 mL
Catalog # 1.09204.0100
Flammable
Toxic
Health
Supplier:   Merck
Giemsa's stain solution for microscopy, Sigma-Aldrich®
Catalog # 1.09204.0100
Supplier:  Merck
CAS Number:  51811-82-6
Giemsa's stain solution for microscopy, Sigma-Aldrich®
Giemsa's stain solution for microscopy, Sigma-Aldrich®

Glass bottle, 100 mL

CAS Number:  51811-82-6
Price
41,20 €
Log in for availability
1 * 100 mL
Product Details & Documents

The Giemsa’s Azure Eosin Methylene Blue solution can be used for the staining of blood and bone marrow smears, paraffin sections and clinical-cytological specimens.

Giemsa staining is one of the standard procedures in histology, demonstrating various cells with their morphological features better than with H&E stain. The Giemsa stain can also be used to detect Helicobacter pylori in gastric tissue biopsies. The specimen material will show cell nuclei stained in various shades of blue, while other structures will appear in various red types. It is an IVD product and CE registered.

Synonyms:  Azure mixture sicc. Giemsa stain, Azur-Eosin-Methylene blue acc. Giemsa, Giemsa's azur-eosin-methylene blue
  • CAS Number:51811-82-6
  • MDL Number:MFCD00081642
  • Boiling Pt:>65 °C (1013 hPa)
  • Density:0.99 g/cm3 (20 °C)
  • Storage Temperature:Ambient
  • ADR:3 (6.1),II
  •  :M-Clarity™ quality level = MQ300
  • Flash Pt:18 °C
  • UN:1230
Specifications

Specification Test Results

  • Suitability for microscopy:
    passes test
  • Erythrocytes:
    reddish-brownish
  • Nuclei:
    violet
  • Eosinophilic granules:
    reddish to red-brown
  • Neutrophilic granules:
    light violet
  • Lymphocyte cytoplasm:
    Blue
  • Monocyte plasma:
    grey-blue
  • Thrombocytes:
    violet
  • Basophilic granules:
    dark violet
  • Blood parasite nuclei:
    light red
  • Nuclei (Tissue section):
    blue to dark blue
  • Cells:
    blue to dark blue
  • Collagen:
    pale blue
  • Osteoid:
    pale blue
  • Eosinophilic granula (Tissue section):
    red
  • Acidic mucopolysaccharides:
    reddish-violet
  • Mast cell granules:
    reddish-violet
  • Cartilage matrix:
    reddish-violet
  • Acidophilic materials:
    orange-red
  • Helicobacter pylori:
    blue to dark blue

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 mL

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.