Flasks, round bottom, with standard ground joint
50 ml, 14/23 NS, 90 ± 6 mm, 51 mm
201-1303
DURAN®, borosilicate glass 3.3, clear.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
With white, matt labelling area.
DIN 12348
: DIN EN ISO 4797
- Ext.Ø:51 mm
- Socket size:14/23 NS
- Capacity:50 ml
- Height:90 ± 6 mm
- Seq:05



