Order Entry
France
Orders LinkContactUsLinkComponent
Nitromethane 96% for spectroscopy

Nitromethane 96% for spectroscopy

Glass bottle, 2,5 L
Catalog # ACRO167860025
Flammable
Irritant
Health
Nitromethane 96% for spectroscopy
Catalog # ACRO167860025
CAS Number:  75-52-5
Nitromethane 96% for spectroscopy
Nitromethane 96% for spectroscopy

Glass bottle, 2,5 L

CAS Number:  75-52-5
Price
719,00 €
Log in for availability
1 * 2,5 L
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
  • CAS Number:75-52-5
  • Formula:CH₃NO₂
  • MDL Number:MFCD00007400
  • Molecular Weight:61.04 g/mol
  • Merck Index:13,06644
  • Boiling Pt:101.2 °C (1013 hPa)
  • Melting Pt:–29 °C
  • Density:1.12 g/cm³
  • Flash Pt:35 °C
  • Storage Temperature:Ambient
  • EINECS:200-876-6
  • UN:1261
  • ADR:3,II

Documents

Specifications

Specification Test Results

  • Appearance:
    Clear liquid
  • Water:
    ≤0.03 %
  • GC:
    ≥95 %
  • UV:
    at 380 nm A: ≤1
  • Color scale:
    ≤10 APHA

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    2,5 L

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.