GLOVES PROFOOD : 400 SERIES : SINGLE-USE GLOVES NATURAL COLOUR AMBIDEXTROUS LATEX. POWDER FREE. TEXTURED FINISH.THICKNESS 0.12 MM. LENGTH 240 MM.100 PIECES IN A DISPENSER. 10 DISPENSERS IN A CARTON. SIZE 9-10.5 1 * 100 items
Catalog # 112-3547
Supplier: Ansell
Frequently Bought Together