Order Entry
France
Orders LinkContactUsLinkComponent
(±)-2-Hexanol 99%

(±)-2-Hexanol 99%

Glass bottle for solids, 50 g
Catalog # ACRO156290500
Flammable
Irritant
(±)-2-Hexanol 99%
Catalog # ACRO156290500
CAS Number:  626-93-7
(±)-2-Hexanol 99%
(±)-2-Hexanol 99%

Glass bottle for solids, 50 g

CAS Number:  626-93-7
Price
161,00 €
Log in for availability
1 * 50 g
Product Details & Documents
Synonyms:  (±)-Methyl butyl carbinol, (±)-Butyl methyl carbinol, (±)-2-Hydroxy hexane
  • CAS Number:626-93-7
  • Formula:C₆H₁₄O
  • MDL Number:MFCD00004585
  • Molecular Weight:102.18 g/mol
  • Boiling Pt:139 °C (760 mmHg)
  • Melting Pt:–23 °C
  • Density:0.81 g/cm³
  • Flash Pt:41 °C
  • Storage Temperature:Ambient
  • EINECS:210-971-4
  • UN:2282
  • ADR:3,III

Documents

Specifications

Specification Test Results

  • Appearance:
    Clear colorless Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4125 to 1.4155 (20°C, 589 nm)
  • GC:
    ≥98.5 %

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    50 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.