N-Bromosuccinimide 99%
Glass bottle for solids, 100 g
Supplier: Thermo Fisher Scientific
N-Bromosuccinimide 99%
Catalog # ACRO107451000
Supplier: Thermo Fisher Scientific
CAS Number: 128-08-5
Jump to:
Product Details & Documents
Synonyms: NBS, 1-Bromopyrrolidine-2,5-dione
- CAS Number:128-08-5
- Formula:C₄H₄BrNO₂
- MDL Number:MFCD00005510
- Molecular Weight:177.99 g/mol
- Merck Index:13,01423
- Boiling Pt:339 °C (1013 hPa)
- Melting Pt:175…178 °C (dec.)
- Density:2,098 g/cm³ (20 °C)
- Flash Pt:180 °C
- Storage Temperature:Refrigerator
- EINECS:204-877-2
- UN:3084
- ADR:8,II
Specifications
Specification Test Results
- Appearance:White to light yellow Crystalline powder
- Infrared spectrum:Conforms
- Titration Iodometric:98.5 to 101.5 % (on dry substance)
- Loss on drying:≤1 %
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g



