Order Entry
France
Orders LinkContactUsLinkComponent
Lithium chloride 99+% ACS

Lithium chloride 99+% ACS

Plastic bottle, 500 g
Catalog # ACRO413275000
Irritant
Lithium chloride 99+% ACS
Catalog # ACRO413275000
CAS Number:  7447-41-8
Lithium chloride 99+% ACS
Lithium chloride 99+% ACS

Plastic bottle, 500 g

CAS Number:  7447-41-8
Price
275,00 €
Log in for availability
1 * 500 g
Product Details & Documents
  • CAS Number:7447-41-8
  • Formula:LiCl
  • MDL Number:MFCD00011078
  • Molecular Weight:42.39 g/mol
  • Merck Index:13,05550
  • Boiling Pt:1382 °C (760 mmHg)
  • Melting Pt:605 °C
  • Density:2.06 g/cm³
  • Storage Temperature:Ambient
  • EINECS:231-212-3

Documents

Specifications

Specification Test Results

  • Appearance:
    White Crystalline powder or crystals
  • Titration Argentometric:
    ≥99 %
  • Loss on drying:
    ≤1.0 % (1 g, 105°C, 2 hrs)
  • Sulfate (SO4):
    ≤100 ppm
  • Barium (Ba):
    ≤30 ppm
  • Calcium (Ca):
    ≤100 ppm
  • Iron (Fe):
    ≤10 ppm
  • Potassium (K):
    ≤100 ppm
  • Sodium (Na):
    ≤2000 ppm
  • Heavy metals (ICP-OES):
    ≤20 ppm
  • Titrable base:
    ≤0.008 meq/g
  • Insoluble matter:
    ≤0.01 %

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    500 g

Specifications