Order Entry
France
Orders LinkContactUsLinkComponent
 

(S)-(-)-2-Methoxypropionic acid 98%

Glass bottle, 2,5 g
Catalog # ACRO331720025
Corrosive
(S)-(-)-2-Methoxypropionic acid 98%
Catalog # ACRO331720025
CAS Number:  23953-00-6
(S)-(-)-2-Methoxypropionic acid 98%
(S)-(-)-2-Methoxypropionic acid 98%

Glass bottle, 2,5 g

CAS Number:  23953-00-6
Price
564,00 €
Log in for availability
1 * 2,5 g
Product Details & Documents

Appearance: Clear colorless to light yellow Liquid

Synonyms:  (S)-2-Methoxypropanoic acid
  • CAS Number:23953-00-6
  • Formula:C₄H₈O₃
  • MDL Number:MFCD01632589
  • Molecular Weight:104.105 g/mol
  • Boiling Pt:99 °C (20 mmHg)
  • Flash Pt:87 °C
  • Storage Temperature:Ambient
  • UN:3265
  • ADR:8,III

 

Specifications

Specification Test Results

  • Appearance:
    Clear colorless to light yellow Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4132 to 1.4152 (20°C, 589 nm)
  • GC:
    ≥97.5 %
  • Specific optical rotation:
    -70° to -80° (20°C, 589 nm) (neat)

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    2,5 g

Specifications