Accessories for T 10 basic, T 18 basic, T 25 digital and T 50 basic
Noix de serrage, support Ø 25 - 36 mm, bras d'extension Ø 5 - 21 mm
Catalog # 431-0004
Supplier: IKA
Accessories for T 10 basic, T 18 basic, T 25 digital and T 50 basic
Catalog # 431-0004
Supplier: IKA
Jump to:
Product Details & Documents
Specifications
- Désignation:Noix de serrage, support Ø 25 - 36 mm, bras d'extension Ø 5 - 21 mm
- Seq:102
Accessories
Product Information
- Catalog #DésignationAvailabilityPrice
- 431-2944Attache souple de sécurité, pour tige Ø 25 - 36 mm et flacon Ø 40 - 300 mmLog in for availability1 * 1 Pce - 517,00 €