Order Entry
France
ContactUsLinkComponent
Cylinder measuring, capacity 10ml, division 0.2ml, polymethylpentene transparent polymer, tall shape, class B, crystal-clear, with e levated scale and main point ring graduation, hexagonal base. 1 * 12 items
Catalog # HIRS7510160
Supplier:  HIRSCHMANN
undefined
Cylinder measuring, capacity 10ml, division 0.2ml, polymethylpentene transparent polymer, tall shape, class B, crystal-clear, with e levated scale and main point ring graduation, hexagonal base. 1 * 12 items
Catalog # HIRS7510160
Supplier:  HIRSCHMANN
Frequently Bought Together