Order Entry
France
Orders LinkContactUsLinkComponent
 

Fixed storage, cabinets and display cases

Standard closet with solid doors, 1200x420x2000 mm
  132-2726
 :   EQUIP LABO
Fixed storage, cabinets and display cases
  132-2726
Fixed storage, cabinets and display cases
Fixed storage, cabinets and display cases

Standard closet with solid doors, 1200x420x2000 mm

 
940,00 €
   
1 * 1 Pce
 

Cabinets in melamine with a super-agglomerate face. Grey finish with white handles. Metal base with adjusting cylinder.

Display cabinet (delivered assembled in 2 sections):
Lower section: solid doors, an adjustable shelf (lower section height: 900 mm)
Upper section: sliding glass doors, two adjustable shelves (upper section height: 1100 mm)
Maximum load per shelf: 15 kg

Cabinet with solid doors (self-assembly):
4 shelves
Maximum load per shelf: 30 kg

Cabinet with solid doors (delivered assembled):
2 vertical drawers mounted on telescopic arms (maximum load for a drawer: 90 kg), or 15 kg per shelf
2 fixed shelves with raised edges and 4 mobile shelves with raised edges (per drawer)
Upper section must be attached to the wall (mounting point)

 
  • Description:
    Standard closet with solid doors
  • W×D×H:
    1200x420x2000 mm