Order Entry
France
Orders LinkContactUsLinkComponent
Taps

Taps

Straight supply hose, length 80 mm, On table
Catalog # 132-2706
Supplier:   EQUIP LABO
Taps
Catalog # 132-2706
Supplier:  EQUIP LABO
Taps
Taps

Straight supply hose, length 80 mm, On table

Price
14,80 €
Log in for availability
1 * 1 Pce
Product Details & Documents

The taps are of 3 types:

  • Taps for hard and demineralised water
  • Taps for combustible gases
  • Taps for air, compressed gas and vacuum

Straight hose length 80 cm, exterior stainless steel braid, interior neoprene sheath. Suitable for taps on the table-top (for cold water, hot water, compressed air). Outlet by 15/21 female nut for connection on 15/21 (¹/₂") male standby gas feed.

Elbow hose with same characteristics as straight hoses but adapted to plumbing fittings on splash back (for cold water, hot water, compressed air).

Ordering information: Different sockets (individual and in a row) are available on request.

Caution: * All of these valves are available in a counter-top version.

Specifications
  • Description:
    Straight supply hose, length 80 mm, On table
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.