BUFFER PH 7 GEEL 20-25 C 1 * 500 mL
Catalog # REGE10705CTT
Supplier: REAGECON
Frequently Bought Together
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
BUFFER PH 4 ROOD 20-25 C 1 * 500 mL
Supplier: REAGECON
VWR®, Nitrile Gloves
Supplier: VWR Collection
Sand, TECHNICAL, industrial, washed
Supplier: VWR Chemicals
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
Samplers for liquids, TeleScoop
Supplier: BURKLE
L(+)-Tartaric acid ≥99.5%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Hydrochloric acid 0.1 mol/l (0.1 N), AVS TITRINORM Reag. Ph. Eur. volumetric solution
Supplier: VWR Chemicals
BUFFER SOLUTION PH10 TWIN NECK 1 * 500 mL
Supplier: REAGECON
VWR®, Serological Pipettes, Open End
Supplier: VWR Collection
Bottles, narrow neck, round, PP, with screw caps, Azlon®
Supplier: SciLabware