Order Entry
France
Orders LinkContactUsLinkComponent
Dynamometers
Dynamometers
0 - 100 N
Catalog # 761-0023
Supplier:   BREUKHOVEN
Dynamometers
Catalog # 761-0023
Supplier:  BREUKHOVEN
Dynamometers
Dynamometers
0 - 100 N
Price
Inquire For Price
Log in for availability
1 * 1 Pce
Product Details & Documents
Robust dynamometers.
  • Transparent body with easy to read scale
Specifications
  • Range:
    0 - 100 N