BASIC Plus Laboratory Coats
Blue, XXL
Catalog # KIMW10034
New Product
Supplier: KIMBERLY CLARK
Jump to:
Product Details & Documents
These coats are made of a protective three-layer SMS fabric which is resistant to blood strike-through. Recommended for areas where there is the potential for light fluid contact.
- Knit collar and cuffs
- Five snap buttons
- Three reinforced pockets
- With size/name label
- Neck loop for hanging
Specifications
- Clothing Size:XXL
- Colour:Blue
- Cat. No.:KIMW10034
- Garment application:Laboratory|Clinical
- Gender:Unisex
- Pockets:With pockets
- Closure type:Press studs / snaps
- Environmentally Preferable:Waste Reducing