Order Entry
France
Orders LinkContactUsLinkComponent
Fmoc-D-Tle-OH

Fmoc-D-Tle-OH

Bottle
Catalog # APOSBICR125-500G
New Product
Supplier:   Apollo Scientific
Fmoc-D-Tle-OH
Catalog # APOSBICR125-500G
Supplier:  Apollo Scientific
New Product
CAS Number:  198543-64-5
Fmoc-D-Tle-OH

Bottle

CAS Number:  198543-64-5
Price
1 770,00 €
Log in for availability
1 * 500 g
Product Details & Documents
Synonyms:  Fmoc-tBu-D-Gly-OH
  • CAS Number:198543-64-5
  • Formula:C₂₁H₂₃NO₄
  • MDL Number:MFCD00065650
  • Molecular Weight:353.418 g/mol
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.