Order Entry
France
Orders LinkContactUsLinkComponent
2-Iodoadenosine

2-Iodoadenosine

Bottle
Catalog # APOSOR310610-25G
New Product
Supplier:   Apollo Scientific
2-Iodoadenosine
Catalog # APOSOR310610-25G
Supplier:  Apollo Scientific
New Product
CAS Number:  35109-88-7
2-Iodoadenosine

Bottle

CAS Number:  35109-88-7
Price
2 220,00 €
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  (2R,3R,4S,5R)-2-(6-Amino-2-iodo-9H-purin-9-yl)-5-(hydroxymethyl)tetrahydrofuran-3,4-diol
  • CAS Number:35109-88-7
  • Formula:C₁₀H₁₂IN₅O₄
  • MDL Number:MFCD01320413
  • Molecular Weight:393.14 g/mol
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle

Specifications