Order Entry
France
Orders LinkContactUsLinkComponent
ε-Caprolactam 99%

ε-Caprolactam 99%

Bottle
Catalog # APOSOR42190-25KG
Irritant
New Product
Supplier:   Apollo Scientific
ε-Caprolactam 99%
Catalog # APOSOR42190-25KG
Supplier:  Apollo Scientific
New Product
CAS Number:  105-60-2
ε-Caprolactam 99%

Bottle

CAS Number:  105-60-2
Price
2 540,00 €
Log in for availability
1 * 25 kg
Product Details & Documents
Synonyms:  Azepan-2-one, 2-Oxohexamethyleneimine, 6-Aminocaproic acid lactam
  • CAS Number:105-60-2
  • Formula:C₆H₁₁NO
  • MDL Number:MFCD00006936
  • Molecular Weight:113.16 g/mol
  • Boiling Pt:267 °C
  • Melting Pt:70 °C
  • Flash Pt:130 °C
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.