Order Entry
France
Orders LinkContactUsLinkComponent
ε-Caprolactam 99%

ε-Caprolactam 99%

Bottle
Catalog # APOSOR42190-500G
Irritant
New Product
Supplier:   Apollo Scientific
ε-Caprolactam 99%
Catalog # APOSOR42190-500G
Supplier:  Apollo Scientific
New Product
CAS Number:  105-60-2
ε-Caprolactam 99%

Bottle

CAS Number:  105-60-2
Price
57,90 €
Log in for availability
1 * 500 g
Product Details & Documents
Synonyms:  Azepan-2-one, 2-Oxohexamethyleneimine, 6-Aminocaproic acid lactam
  • CAS Number:105-60-2
  • Formula:C₆H₁₁NO
  • MDL Number:MFCD00006936
  • Molecular Weight:113.16 g/mol
  • Boiling Pt:267 °C
  • Melting Pt:70 °C
  • Flash Pt:130 °C
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle

Specifications