Order Entry
France
Orders LinkContactUsLinkComponent
TD139 ≥95%

TD139 ≥95%

Bottle
Catalog # APOSBUP06798-50MG
New Product
Supplier:   Apollo Scientific
TD139 ≥95%
Catalog # APOSBUP06798-50MG
Supplier:  Apollo Scientific
New Product
CAS Number:  1450824-22-2
TD139 ≥95%

Bottle

CAS Number:  1450824-22-2
Price
990,00 €
Log in for availability
1 * 50 mg
Product Details & Documents

Usage: Biochemical/life science applications.

Synonyms:  TD139, (2R,2'R,3R,3'R,4S,4'S,5R,5'R,6S,6'S)-6,6'-Thiobis(4-(4-(3-fluorophenyl)-1H-1,2,3-triazol-1-yl)-2-(hydroxymethyl)tetrahydro-2H-pyran-3,5-diol), (2S,3R,4S,5R,6R)-4-[4-(3-fluorophenyl)triazol-1-yl]-2-[(2S,3R,4S,5R,6R)-4-[4-(3-fluorophenyl)triazol-1-yl]-3,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl]sulphanyl-6-(hydroxymethyl)oxane-3,5-diol
  • CAS Number:1450824-22-2
  • Formula:C₂₈H₃₀F₂N₆O₈S
  • Molecular Weight:648,64 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications