Order Entry
France
Orders LinkContactUsLinkComponent
 

Amonafide 98%

Bottle
Catalog # APOSOR1023061-1G
New Product
Supplier:   Apollo Scientific
Amonafide 98%
Catalog # APOSOR1023061-1G
Supplier:  Apollo Scientific
New Product
CAS Number:  69408-81-7
Amonafide 98%

Bottle

CAS Number:  69408-81-7
Price
911,00 €
Log in for availability
1 * 1 g
Product Details & Documents

Usage: Organic building block/reagent.

Synonyms:  Amonafide
  • CAS Number:69408-81-7
  • Formula:C₁₆H₁₇N₃O₂
  • Molecular Weight:283,33 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications