Order Entry
France
Orders LinkContactUsLinkComponent
Accessories for Inverted Microscope, Axiovert 5
Accessories for Inverted Microscope, Axiovert 5
Mechanical stage 130×85 R/L for Axiovert 5 and Axio Vert.A1, manual 3-plate mechanical stage, for mounting frames K 160×110 mm, max. 3 kg sample load
Catalog # ZEIS4320439000000
New Product
Supplier:   ZEISS
Accessories for Inverted Microscope, Axiovert 5
Catalog # ZEIS4320439000000
Supplier:  ZEISS
New Product
Accessories for Inverted Microscope, Axiovert 5
Mechanical stage 130×85 R/L for Axiovert 5 and Axio Vert.A1, manual 3-plate mechanical stage, for mounting frames K 160×110 mm, max. 3 kg sample load
Price
Inquire For Price
Log in for availability
1 * 1 Pce
Product Details & Documents
Specifications
  • Description:
    Mechanical stage 130×85 R/L for Axiovert 5 and Axio Vert.A1, manual 3-plate mechanical stage, for mounting frames K 160×110 mm, max. 3 kg sample load