Order Entry
France
ContactUsLinkComponent
Microfil® Silicon Tubing
Microfil® Silicon Tubing
  MILXMIACTUB01
 :  Merck
New Product
Microfil® Silicon Tubing
  MILXMIACTUB01
 :  Merck
New Product

 

  • Description:
    Silicon tubing
  • Pk:
    1 unit

 

 

The silicon tubing is an additional accessory of the Microfil® filtration system. The Microfil® filtration system is a simple and reliable solution for routine microbiological testing. The push-fit design of the Microfil® funnels seals tightly to manifold supports without the need for clamps or O-rings. The tube is used to connect the pump to the manifold or filtration apparatus using the Tee connectors. The tubing is autoclavable, 5 m long, with an internal diameter of 9,5 mm.

The silicon tubing is an additional accessory of the Microfil® filtration system used in general microbiological analysis of beer, bottled water, pharmaceutical products analysis, environmental analysis, bioburden testing, and water monitoring.

Input: Sample type beverage(s), sample type food(s).
Application: Bioburden testing, clinical, environmental, food and beverages, industrial qc, water monitoring.
Compatibility: For use with Microfil® filtration system.
Shipped in: Ambient.