Order Entry
France
ContactUsLinkComponent
NANOCOLOR® Photometric Tube Tests, Chlorine dioxide
NANOCOLOR® Photometric Tube Tests, Chlorine dioxide
Catalog # MANA985018
Supplier:  MACHEREY-NAGEL
CAS Number:  
undefined
NANOCOLOR® Photometric Tube Tests, Chlorine dioxide
Catalog # MANA985018
Supplier:  MACHEREY-NAGEL
CAS Number:  

Specifications

  • Chemical method:
    DPD
  • Pk:
    1 KIT
  • Description:
    NANOCOLOR® Chlorine dioxide 5, test no. 0-18
  • Dimensions:
    160×110×119 mm (Including packaging)
  • Model:
    MANA985018
  • Platform:
    NANOCOLOR® tube tests
  • Storage temperature:
    15...25 °C
  • Shelf life (months):
    12
  • Weight:
    340,7 g (Including packaging)
  • For unit:
    UV/VIS II;VIS II;500 D;PF-12Plus;Advance
  • Tests per kit:
    20
  • Range:
    0,15 - 5,00 mg/L

Specifications

About this item

Photometric test kit for the determination of chlorine dioxide in water samples.

  • Measurement directly in the test tube, saving time and reducing exposure to chemical reagents
  • Bar code cuvette identification for easy test call up in NANOCOLOR® spectrophotometers
  • Precisely pre-dosed reagents with constant high quality from batch to batch
  • Colored pictogram instructions and easy handling of the tubes
  • 16 mm (OD) tubes for easy pipetting

The test kit is evaluated in 16 mm (AD) round cells. It is pre-programmed on the NANOCOLOR® photometers. Programming data for other spectrophotometers are available.

Sea water analysis: Yes

Hazardous material: Yes

Remark: Measuring range on NANOCOLOR VIS II

Sea water analysis: No

Hazardous material: Yes

Remark: Measuring range on NANOCOLOR VIS II

Sludge reagent: No

Digestion: No

NanOx metal: No

NanOx: No

Delivery:

Rugged box with 20 test tubes. Sufficient for 20 tests.